missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SPAG8 Antibody (2F12), Novus Biologicals™

Product Code. 18346539 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346539 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346539 Supplier Novus Biologicals™ Supplier No. H00026206M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SPAG8 Monoclonal antibody specifically detects SPAG8 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPAG8
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2F12
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH19247
Gene Alias BS-84, HSD-1MGC26201, hSMP-1, SMP1, SMP-1, SPAG3, sperm associated antigen 8, Sperm membrane protein 1, Sperm membrane protein BS-84, sperm-associated antigen 8
Host Species Mouse
Immunogen SPAG8 (AAH19247, 1 a.a. ∽ 501 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. METNGSTEGSRSRSRSLDIQPSSEGLGPTSEPFPSSDDSPRSALAAATAAAAAAASAAAATAAFTTAKAAALSTKTPAPCSEFMEPSSDPSLLGEPCAGPGFTHNIAHGSLGFEPVYVSCIAQDTCTTTDHSSNPGPVPGSSSGPVLGSSSGAGHGSGSGSGPGCGSVPGSGSGPGPGSGPGSGPGHGSGSHPGPASGPGPDTGPDSELSPCIPPGFRNLVADRVLNYTSWSQHCPWEPQKQPPWEFLQVLEPGARGLWKPPDIKGKLMVCYETLPRGQCLLYNWEEERATNHLDQVPSMQDGSESFFFRHGHRGLLTMQLKSPMPSSTTQKDSYQPPGNVYWPLRGKREAMLEMLLQHQICKEVQAEQEPTRKLFEVESVTHHDYRMELAQAGTPAPTKPHDYRQEQPETFWIQRAPQLPTWWPLPTQVPAAEDYLTWKEWGFTGVQEVLSALLRATPGEYSVNICGMNEHPVCSRTWTNRLCHQEMGSKKTVTQEDRGW
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26206
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.