missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPAG7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565719
This item is not returnable.
View return policy
Description
Immunogen sequence: DDDCRYVMIFK KEFAPSDEEL DSYRRGEEWD PQKAEEKRKL KELAQRQEEE AAQQGPVVVS PASDYKDKYS HLIGKGAAKD AAHMLQANKT YGC Highest antigen sequence identity to the following orthologs - mouse 97%, rat 96%.
SPAG7 gene ontology annotations related to this gene include nucleic acid binding.
Specifications
| SPAG7 | |
| Polyclonal | |
| Unconjugated | |
| SPAG7 | |
| 5730443G10; ACRP; FSA-1; Fsa1l; hypothetical protein LOC515086; similar to sperm associated antigen 7; SPAG7; sperm associated antigen 7; sperm-associated antigen 7; Sperm-associated antigen 7 homolog; zgc:66397 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9552 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75391 | |
| SPAG7 | |
| Recombinant Human SPAG7. Recombinant protein control fragment (Product #RP-104943). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction