missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPAG4L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100185
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SPAG4L.
The function of this protein remains unknown.
Specifications
| SPAG4L | |
| Polyclonal | |
| Unconjugated | |
| SUN5 | |
| 1700021O15Rik; dJ726C3.1; Sad1 and UNC84 domain containing 5; sad1 and UNC84 domain-containing protein 5; SPAG4L; SPAG4L-2; Sperm-associated antigen 4-like protein; SRG4; SUN domain-containing protein 5; SUN5; testis and spermatogenesis related gene 4; testis and spermatogenesis-related gene 4 protein; TSARG4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140732, 76407 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8TC36, Q9DA32 | |
| SUN5 | |
| A synthesized peptide derived from human SUN5(Accession Q8TC36), corresponding to amino acid residues A266-F316. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction