missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG4L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13399-25ul
This item is not returnable.
View return policy
Description
SPAG4L Polyclonal specifically detects SPAG4L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SPAG4L | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| dJ726C3.1, MGC33594, Sad1 and UNC84 domain containing 5, SPAG4L, Sperm-associated antigen 4-like protein, SUN domain-containing protein 5, testis and spermatogenesis related gene 4, Testis and spermatogenesis-related gene 4 protein, TSARG4sperm associated antigen 4-like | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 140732 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SUN5 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: MPRSSRSPGDPGALLEDVAHNPRPRRIAQRGRNTSRMAEDTSPNMNDNILLPVRNNDQA | |
| 25ul | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction