missing translation for 'onlineSavingsMsg'
Learn More

SPAG4 Antibody (OTI3E9) [DyLight 680], Novus Biologicals™

Product Code. 30688764 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688764 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688764 Supplier Novus Biologicals Supplier No. NBP344837FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SPAG4 Monoclonal antibody specifically detects SPAG4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPAG4
Applications Western Blot
Classification Monoclonal
Clone OTI3E9
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias acrosomal protein ACR55, CT127, Outer dense fiber-associated protein SPAG4, Sad1 and UNC84 domain containing 4, sperm associated antigen 4, sperm tail protein, sperm-associated antigen 4 protein, SUN domain-containing protein 4, SUN4cancer/testis antigen 127
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 191-279 of human SPAG4 (NP_003107) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6676
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.