missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPAG16 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142541
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize isoform 1 and 2 (NP_078808.3; NP_001020607.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Cilia and flagella are comprised of a microtubular backbone, the axoneme, which is organized by the basal body and surrounded by plasma membrane. SPAG16 encodes 2 major proteins that associate with the axoneme of sperm tail and the nucleus of postmeiotic germ cells, respectively.
Specifications
| SPAG16 | |
| Polyclonal | |
| Unconjugated | |
| SPAG16 | |
| 4921511D23Rik; 4930524F24Rik; 4930585K05Rik; AV261009; PF20; Pf20 protein homolog; RGD1565062; SPAG16; sperm associated antigen 16; sperm-associated antigen 16 protein; sperm-associated WD repeat protein; spg16; WD repeat domain 29; WDR29 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 66722, 79582 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8K450, Q8N0X2 | |
| SPAG16 | |
| Peptide with sequence C-KGVTELRTVGNVPD. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction