missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPAG11B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544106
This item is not returnable.
View return policy
Description
Peptide sequence: MRQRLLPSVT SLLLVALLFP GSSQARHVNH SATEALGELR ERAPGQGTNG Sequence homology: Human: 100%.
SPAG11B is one of several androgen-dependent, epididymis-specific secretory proteins. The specific functions of these proteins have not been determined, but they are thought to be involved in sperm maturation. Some of the isoforms contain regions of similarity to beta-defensins, a family of antimicrobial peptides.This gene encodes several androgen-dependent, epididymis-specific secretory proteins. The specific functions of these proteins have not been determined, but they are thought to be involved in sperm maturation. Some of the isoforms contain regions of similarity to beta-defensins, a family of antimicrobial peptides. The gene is located on chromosome 8p23 near the defensin gene cluster. Alternative splicing of this gene results in seven transcript variants encoding different isoforms. Two different N-terminal and five different C-terminal protein sequences are encoded by the splice variants. Two additional variants have been described, but their full length sequences have not been determined.
Specifications
| SPAG11B | |
| Polyclonal | |
| Unconjugated | |
| SPAG11B | |
| EDDM2B; EP2; EP2C; EP2D; epididymal protein 2B; HE2; HE2C; Human epididymis-specific protein 2; MGC61846; Protein EP2; SPAG11; SPAG11A; SPAG11B; Sperm antigen HE2; sperm associated antigen 11B; Sperm-associated antigen 11A; Sperm-associated antigen 11B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10407 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q08648 | |
| SPAG11B | |
| Synthetic peptide directed towards the N-terminal of human SPAG11B (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction