missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG1 Antibody (10G1/2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50301
This item is not returnable.
View return policy
Description
SPAG1 Monoclonal antibody specifically detects SPAG1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| SPAG1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| FLJ32920, HSD-3.8sperm-associated antigen 1, Infertility-related sperm protein Spag-1, SP75, sperm associated antigen 1, tetratricopeptide repeat-containing protein, TPIS, TPR-containing protein involved in spermatogenesis | |
| Synthetic peptide SPA-3 (sequence designed to avoid TPR motifs - KTAPFNKEKERRKIEIQEVNE) | |
| 0.1 mL | |
| Cancer, Developmental Biology | |
| 6674 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| 10G1/2 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction