Learn More
Hamilton™ Spacer
Click to view available options
Unit Size:
Each
Description
- Also provides a convenient handhold when inserting the needle
- 51mm
Specifications
Specifications
| Description | Results in 19mm exposed needle length |
| Product Type | Needle Spacer |
| For Use With (Equipment) | 7000 Series Microliter Syringes |
Product Content Correction
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu