missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPACA9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569983
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
SPACA9 (Sperm Acrosome Associated 9) is a Protein Coding gene.
Specifications
| SPACA9 | |
| Polyclonal | |
| Unconjugated | |
| SPACA9 | |
| 1700026L06Rik; 80-kDa SOGA fragment; Acrosome and sperm tail protein; C11H9orf9; C20orf117; C9H9orf9; C9orf9; ci009; C-terminal 80 kDa form; KIAA0889; MAST; N-terminal form; Protein SOGA1; RIKEN cDNA 1700026L06 gene; Rsb66; Rsb66 homolog; Rsb-66 protein; SOGA; SOGA family member 1; SOGA1; SPACA9; sperm acrosome associated 9; sperm acrosome-associated protein 9; Suppressor of glucose by autophagy; suppressor of glucose from autophagy; suppressor of glucose, autophagy associated 1; suppressor of glucose, autophagy-associated protein 1; uncharacterized protein C9orf9 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 69987 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7TPM5 | |
| SPACA9 | |
| synthetic peptide directed towards the C-terminal region of mouse 1700026L06Rik. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction