missing translation for 'onlineSavingsMsg'
Learn More

SPACA9 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18419476 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419476 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419476 Supplier Novus Biologicals Supplier No. NBP282011AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPACA9 Polyclonal antibody specifically detects SPACA9 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPACA9
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias C20orf117, chromosome 20 open reading frame 117, dJ132F21.1, FLJ44670, hypothetical protein LOC140710, KIAA0889, SOGA, suppressor of glucose, autophagy associated 1
Host Species Rabbit
Immunogen SPACA9 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human SPACA9. The immunogen is located within the last 50 amino acids of SPACA9 . Amino Acid Squence: RLHPSESSWGGEERA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 140710
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.