missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Positive Samples: Rat testis, Rat brain Immunogen sequence: KRFTRSDELE RHVRTHTREK KFTCLLCSKR FTRSDHLSKH QRTH.
SP7 is a C2H2-type zinc finger transcription factor of the SP gene family and a putative master regulator of bone cell differentiation (Gao et al., 2004 [PubMed 15474293]).
Specifications
Specifications
| Antigen | SP7 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.55 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | SP7 |
| Gene Accession No. | Q8TDD2, Q8VI67 |
| Gene Alias | 6430578P22Rik; C22; MGC126598; OI11; OI12; osterix; OSX; Sp7; Sp7 transcription factor; Sp7 transcription factor 7; trans-acting transcription factor 7; transcription factor Sp7; Zinc finger protein osterix |
| Gene Symbols | SP7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
