missing translation for 'onlineSavingsMsg'
Learn More

SP600125, MedChemExpress

Product Code. 16471026
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471026 200 mg 200mg
16434948 10 mg 10mg
16415726 100 mg 100mg
16401197 50 mg 50mg
16461626 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16471026 Supplier Medchem Express Supplier No. HY12041
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SP600125 is an orally active, reversible, and ATP-competitive JNK inhibitor with IC50s of 40, 40 and 90 nM for JNK1, JNK2 and JNK3, respectively. SP600125 is a potent ferroptosis inhibitor. SP600125 inhibits autophagy and activates apoptosis.

SP600125 is an orally active, reversible, and ATP-competitive JNK inhibitor with IC50s of 40, 40 and 90 nM for JNK1, JNK2 and JNK3, respectively. SP600125 is a potent ferroptosis inhibitor. SP600125 inhibits autophagy and activates apoptosis.

Specifications

Chemical Name or Material SP600125
CAS 129-56-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H8N2O
Quantity 200 mg
Solubility Information DMSO : 12.5 mg/mL (56.76 mM; Need ultrasonic)
SMILES O=C1C2=C3C(NN=C3C4=C1C=CC=C4)=CC=C2
Molecular Weight (g/mol) 220.23
Formula Weight 220.23
Percent Purity 96.79%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.