missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies SP600125

Product Code. 17120102
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17120102 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17120102 Supplier STEMCELL Technologies Supplier No. 72642
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

JNK pathway inhibitor; Inhibits JNK1, JNK2, and JNK3

  • SP600125 is a potent and reversible inhibitor of JNK1-3 (IC50 = 0.11 μM) (Bennett et al.) It is cell-permeable and dose-dependently inhibits c-JUN phosphorylation in cells, blocking the expression of COX-2 and TNF-α in monocytes and IL-10, TNF-α, and IFN- in T-cells (Bennett et al.)
  • Direct lineage reprogramming of fibroblasts to mature neurons, in combination with CHIR99021, RepSox, Forskolin, Gö6983, VaLproic Acid and Y-27632 (Hu et al.)
  • Inhibits BMP9-induced osteogenic differentiation in cultured mouse mesenchymal stem cells (MSCs) and in primary bone marrow stromal cells (Zhao et al.)
  • Promotes adipogenic, but represses osteogenic differentiation of human MSCs (Bilkovski et al.; Liu et al.; Qiu et al.; Tominaga et al.)
  • Causes cell death and inhibits neurogenesis when added during early stages of neuronal culture (Tiwari et al.)

Specifications

CAS 129-56-6
Synonym NSC 75890; Pyrazolanthrone; 1PMV
Quantity 5 mg
Molecular Formula C14H8N2O
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.