missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559159
This item is not returnable.
View return policy
Description
Immunogen sequence: NQATGQQQII IDPSQGLVQL QNQPQQLELV TTQLAGNAWQ LVASTPPASK ENNVSQP Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene belongs to a family of Sp1 related genes that encode transcription factors that regulate transcription by binding to consensus GC- and GT-box regulatory elements in target genes. This protein contains a zinc finger DNA-binding domain and several transactivation domains, and has been reported to function as a bifunctional transcription factor that either stimulates or represses the transcription of numerous genes. Transcript variants encoding different isoforms have been described for this gene, and one has been reported to initiate translation from a non-AUG (AUA) start codon. Additional isoforms, resulting from the use of alternate downstream translation initiation sites, have also been noted. A related pseudogene has been identified on chromosome 13.
Specifications
| SP4 | |
| Polyclonal | |
| Unconjugated | |
| Sp4 | |
| 5730497N03Rik; HF1B; HF-1b; HF1-b; HF-1b zinc finger protein; LOW QUALITY PROTEIN: transcription factor Sp4; MGC130008; MGC130009; SP4; Sp4 transcription factor; SPR-1; trans-acting transcription factor 4; transcription factor Sp4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6671 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q02446 | |
| Sp4 | |
| Recombinant protein corresponding to Human SP3/SP4. Recombinant protein control fragment (Product #RP-98389). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction