missing translation for 'onlineSavingsMsg'
Learn More

Sp3 Antibody (2F6), Novus Biologicals™

Product Code. 18322609 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322609 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322609 Supplier Novus Biologicals Supplier No. H00006670M08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sp3 Monoclonal antibody specifically detects Sp3 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sp3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2F6
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003102
Gene Alias DKFZp686O1631, GC-binding transcription factor Sp3, Sp3 transcription factor, specificity protein 3, SPR2, SPR-2, transcription factor Sp3
Host Species Mouse
Immunogen SP3 (NP_003102, 287 a.a. ~ 430 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TAGINADGHLINTGQAMDSSDNSERTGERVSPDINETNTDTDLFVPTSSSSQLPVTIDSTGILQQNTNSLTTSSGQVHSSDLQGNYIQSPVSEETQAQNIQVSTAQPVVQHLQLQESQQPTSQAQIVQGITPQTIHGVQASGQN*
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6670
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.