missing translation for 'onlineSavingsMsg'
Learn More

SP2509, MedChemExpress

Product Code. 16481477
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481477 50 mg 50mg
16475268 10 mg 10mg
16437987 5 mg 5mg
16415936 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481477 Supplier Medchem Express Supplier No. HY12635
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SP2509 is a potent and selective antagonist of lysine specific demethylase 1 (LSD1) with an IC50 of 13 nM.

SP2509 is a potent and selective antagonist of lysine specific demethylase 1 (LSD1) with an IC50 of 13 nM.

Specifications

Chemical Name or Material SP2509
CAS 1423715-09-6
Color White
Purity Grade Notes Research
Molecular Formula C19H20ClN3O5S
Quantity 50 mg
Solubility Information DMSO : ≥ 33 mg/mL (75.36 mM)
SMILES O=C(N/N=C(C1=CC(Cl)=CC=C1O)\C)C2=CC=CC(S(=O)(N3CCOCC3)=O)=C2
Molecular Weight (g/mol) 437.9
Formula Weight 437.9
Percent Purity 97.72%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.