missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SP2 Polyclonal Antibody
Description
Recommended positive controls: Jurkat, Raji, NCI-H929, A431, HepG2. Predicted reactivity: Mouse (91%), Rat (90%), Rhesus Monkey (100%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the Sp subfamily of Sp/XKLF transcription factors. Sp family proteins are sequence-specific DNA-binding proteins characterized by an amino-terminal trans-activation domain and three carboxy-terminal zinc finger motifs. This protein contains the least conserved DNA-binding domain within the Sp subfamily of proteins, and its DNA sequence specificity differs from the other Sp proteins. It localizes primarily within subnuclear foci associated with the nuclear matrix, and can activate or in some cases repress expression from different promoters.
Specifications
Specifications
| Antigen | SP2 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.43 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | SP2 |
| Gene Accession No. | Q02086 |
| Gene Alias | 4930480I16Rik; Kiaa0048; mKIAA0048; Sp2; Sp2 transcription factor; Trans-acting transcription factor 2; transcription factor Sp2 |
| Gene Symbols | SP2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?