missing translation for 'onlineSavingsMsg'
Learn More

SP187, MedChemExpress

Product Code. 16450726
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16450726

Brand: Medchem Express HYU00160

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress SP187 is a host-targeted iminosugar with activity against filovirus infections in vitro and in vivo. SP187 is active against influenza and dengue in vivo.

SP187 is a host-targeted iminosugar with activity against filovirus infections in vitro and in vivo. SP187 is active against influenza and dengue in vivo.

Specifications

Chemical Name or Material SP187
Physical Form Solid
CAS 615253-61-7
Color White
Quantity 20 mg
Purity Grade Notes Research
Molecular Formula C16H33NO5
For Use With (Application) COVID-19-anti-virus
Synonym MON-​DNJ UV4
Solubility Information DMSO : 100 mg/mL (313.05 mM; Need ultrasonic)
SMILES OC[C@H]1N(C[C@H](O)[C@@H](O)[C@@H]1O)CCCCCCCCCOC
Molecular Weight (g/mol) 319.44
Formula Weight 319.44
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.