missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sp17 Polyclonal Antibody

Rabbit Polyclonal Antibody
£155.00 - £430.00
Specifications
| Antigen | Sp17 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17750834
![]() |
Invitrogen™
PA5119430 |
200 μL |
£430.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17722984
|
Invitrogen™
PA5121584 |
20 μL |
£155.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sp17 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a protein present at the cell surface. The N-terminus has sequence similarity to human cAMP-dependent protein kinase A (PKA) type II alpha regulatory subunit (RIIa) while the C-terminus has an IQ calmodulin-binding motif. The central portion of the protein has carbohydrate binding motifs and likely functions in cell-cell adhesion. The protein was initially characterized by its involvement in the binding of sperm to the zona pellucida of the oocyte. Recent studies indicate that it is also involved in additional cell-cell adhesion functions such as immune cell migration and metastasis. A retrotransposed pseudogene is present on chromosome 10q22.Specifications
| Sp17 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse, Rat | |
| SPA17 | |
| band 34; cancer/testis antigen 22; CT22; SP17; SP17 protein; SP17-1; SPA17; Sperm autoantigenic protein 17; sperm protein 17; Sperm surface protein Sp17; Unknown (protein for MGC:127855); zgc:153721 | |
| SPA17 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q62252 | |
| 20686, 85244 | |
| Recombinant protein Sperm Protein 17. The antigen corresponds to amino acid range 1-149 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title