missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP17 Antibody (OTI2G6), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74307DL594
This item is not returnable.
View return policy
Description
SP17 Monoclonal antibody specifically detects SP17 in Human samples. It is validated for Western Blot
Specifications
| SP17 | |
| Monoclonal | |
| DyLight 594 | |
| Cancer/testis antigen 22, CT22SP17-1, Sp17-1, SP17sperm surface protein Sp17, sperm autoantigenic protein 17Sperm protein 17 | |
| Full length recombinant protein of human SP17 (NP_059121) produced in E.coli. | |
| 0.1 mL | |
| Cancer | |
| 53340 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| OTI2G6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction