missing translation for 'onlineSavingsMsg'
Learn More

SP17 Antibody (3B6), Novus Biologicals™

Product Code. 18321939 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321939 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321939 Supplier Novus Biologicals Supplier No. H00053340M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SP17 Monoclonal antibody specifically detects SP17 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP17
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3B6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_059121
Gene Alias Cancer/testis antigen 22, CT22SP17-1, Sp17-1, SP17sperm surface protein Sp17, sperm autoantigenic protein 17Sperm protein 17
Host Species Mouse
Immunogen SPA17 (NP_059121, 51 a.a. ~ 149 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EKTNFDPAEWGSKVEDRFYNNHAFEEQEPPEKSDPKQEESQISGKEEETSVTILDSSEEDKEKEEVAAVKIQAAFRGHIAREEAKKMKTNSLQNEEKEE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 53340
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.