missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP140L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563192
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KEREERPDIK LSLKQGEVPE SPEA Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 50%.
SP140L (SP140 Nuclear Body Protein Like) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include chromatin binding. An important paralog of this gene is SP140. [GeneCards]Specifications
SP140L | |
Polyclonal | |
Unconjugated | |
SP140L | |
Nuclear body protein SP140-like protein; SP140 nuclear body protein like; SP140 nuclear body protein-like; SP140L; SP140L-1 protein; SP140L-2 protein; SP140L-3 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
93349 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H930 | |
SP140L | |
Recombinant protein corresponding to Human SP140L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only