missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP140 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £484.00
Specifications
| Antigen | SP140 |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667558
|
Novus Biologicals
NBP2-49517-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654918
|
Novus Biologicals
NBP2-49517 |
0.1 mL |
£484.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SP140 Polyclonal antibody specifically detects SP140 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| SP140 | |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Lymphoid-restricted homolog of Sp100, lymphoid-specific SP100 homolog, LYSP100, LYSP100-A, LYSP100-B, MGC126440, Nuclear autoantigen Sp-140, nuclear body protein SP140, SP140 nuclear body protein, Speckled 140 kDa | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LSLLPGEGEEGSDDCSEMCDGEERQEASSSLARRGSVSSELENHPMNEEGESEELASSLLYDNVPGAEQSA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| 11262 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title