missing translation for 'onlineSavingsMsg'
Learn More

SP140 Antibody, Novus Biologicals™

Product Code. 18397119 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397119 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397119 Supplier Novus Biologicals Supplier No. H00011262B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

SP140 Polyclonal antibody specifically detects SP140 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP140
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_001005176.1
Gene Alias Lymphoid-restricted homolog of Sp100, lymphoid-specific SP100 homolog, LYSP100, LYSP100-A, LYSP100-B, MGC126440, Nuclear autoantigen Sp-140, nuclear body protein SP140, SP140 nuclear body protein, Speckled 140 kDa
Host Species Mouse
Immunogen SP140 (NP_001005176.1, 1 a.a. - 172 a.a.) full-length human protein. MAQQGQQGQMASGDSNLNFRMVAEIQNVEGQNLQEQVCPEPIFRFFRENKVEIASAITRPFPFLMGLRDRSFISEQMYEHFQEAFRNLVPVTRVMYCVLSELEKTFGWSHLEALFSRINLMAYPDLNEIYRSFQNENLSSSAVLCQLVSPNKDWRSHEESLAHTGTLRRSCM
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11262
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.