missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Target | SP140 |
| Synonym | Lymphoid-restricted homolog of Sp100, lymphoid-specific SP100 homolog, LYSP100, LYSP100-A, LYSP100-B, MGC126440, Nuclear autoantigen Sp-140, nuclear body protein SP140, SP140 nuclear body protein, Speckled 140 kDa |
| Gene Symbol | SP140 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?