missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP110 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11502-1-AP-20UL
This item is not returnable.
View return policy
Description
Susceptibility to tuberculosis (TB) in mice has recently been attributed to the IPR1 gene. IPR1 is a member of the SP100/SP140 family of nuclear body proteins and encodes a leukocyte-specific nuclear body component. The protein can function as an activator of gene transcription and may serve as a nuclear hormone receptor coactivator. Alternative splicing has been observed for this gene and three transcript variants, encoding distinct isoforms, have been identified. SP110 is the closest homolog of the IPR1 protein in humans. The IPR1/Sp110 gene product might play a role in integrating signals generated by intracellular pathogens with mechanisms controlling innate immunity, cell death, and pathogenesis. IPR1/Sp110 is up-regulated after infection with M. tuberculosis and required by Anaplasma phagocytophilum for infection of human promyelocytic cells. Defects in Sp110 are a cause of severely impaired resistance to infection by M. tuberculosis.Specifications
| SP110 | |
| Polyclonal | |
| Unconjugated | |
| SP110 | |
| IFI41, IFI75, IPR1, SP110, SP110 nuclear body protein, Speckled 110 kDa, VODI | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109032, 3431 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BVK9, Q9HB58 | |
| SP110 | |
| SP110 Fusion Protein Ag2042 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction