missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SP110 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142487
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Additional Information: This antibody is expected to recognize all three reported isoforms (NP_004500.3; NP_004501.3; NP_536349.2). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
Susceptibility to tuberculosis (TB) in mice has recently been attributed to the IPR1 gene. IPR1 is a member of the SP100/SP140 family of nuclear body proteins and encodes a leukocyte-specific nuclear body component. The protein can function as an activator of gene transcription and may serve as a nuclear hormone receptor coactivator. Alternative splicing has been observed for this gene and three transcript variants, encoding distinct isoforms, have been identified. SP110 is the closest homolog of the IPR1 protein in humans. The IPR1/Sp110 gene product might play a role in integrating signals generated by intracellular pathogens with mechanisms controlling innate immunity, cell death, and pathogenesis. IPR1/Sp110 is up-regulated after infection with M. tuberculosis and required by Anaplasma phagocytophilum for infection of human promyelocytic cells. Defects in Sp110 are a cause of severely impaired resistance to infection by M. tuberculosis.
Specifica
| SP110 | |
| Polyclonal | |
| Unconjugated | |
| SP110 | |
| 5031415C07Rik; 52kDa; 5830484A20Rik; ENSMUSG00000075603; IFI41; Ifi75; interferon-induced protein 41, 30kD; interferon-induced protein 41/75; interferon-induced protein 75, 52kD; intracellular pathogen resistance 1; intracellular pathogen resistance protein 1; Ipr1; phosphoprotein 41; phosphoprotein 75; SP110; Sp110 nuclear body protein; speckled 110 kDa; Transcriptional coactivator Sp110; VODI | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 3431 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9HB58 | |
| SP110 | |
| Peptide with sequence C-PQEVSSTPSDKKGKK. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto