missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SP100 Antibody, Novus Biologicals™

Product Code. 18343259 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343259 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343259 Supplier Novus Biologicals™ Supplier No. H00006672D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SP100 Polyclonal antibody specifically detects SP100 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP100
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH11562.1
Gene Alias DKFZp686E07254, FLJ00340, FLJ34579, lysp100b, nuclear antigen Sp100, nuclear autoantigen Sp-100, Nuclear dot-associated Sp100 protein, SP100 nuclear antigen, SP100-HMG nuclear autoantigen, Speckled 100 kDa
Host Species Rabbit
Immunogen SP100 (AAH11562.1, 1 a.a. - 480 a.a.) full-length human protein. MAGGGGDLSTRRLNECISPVANEMNHLPAHSHDLQRMFTEDQGVDDRLLYDIVFKHFKRNKVEISNAIKKTFPFLEGLRDRDLITNKMFEDSQDSCRNLVPVQRVVYNVLSELEKTFNLPVLEALFSDVNMQEYPDLIHIYKGFENVIHDKLPLQESEEEEREERSGLQLSLEQGTGENSFRSLTWPPSGSPSHAGTTPPENGLSEHPCETEQINAKRKDTTSDKDDSLGSQQTNEQCAQKAEPTESCEQIAVQVNNGDAGREMPCPLPCDEESPEAELHNHGIQINSCSVRLVDIKKEKPFSNSKVECQAQARTHHNQASDIIVISSEDSEGSTDVDEPLEVFISAPRSEPVINNDNPLESNDEKEGQEATCSRPQIVPEPMDFRKLSTFRESFKKRVIGQDHDFSESSEEEAPAEASSGALRSKHGEKAPMTSRSTSTWRIPSRKRRFSSSDFSDLSNGEELQETCSSSLRRGSGKED
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6672
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.