Learn More
SP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21962-1-AP-20UL
 View more versions of this product
Product Code. 16891505
  • £135.00 / 20µL
Description
SP1 is a transcription factor that can activate or repress transcription in response to physiological and pathological stimuli. Binds with high affinity to GC-rich motifs and regulates the expression of a large number of genes involved in a variety of processes such as cell growth, apoptosis, differentiation and immune responses. Highly regulated by post-translational modifications (phosphorylations, sumoylation, proteolytic cleavage, glycosylation and acetylation). Binds also the PDGFR-alpha G-box promoter. May have a role in modulating the cellular response to DNA damage. Implicated in chromatin remodeling. Plays a role in the recruitment of SMARCA4/BRG1 on the c-FOS promoter. Plays an essential role in the regulation of FE65 gene expression.
Specifications
Specifications
SP1
Polyclonal
Unconjugated
SP1
SP1, Sp1 transcription factor, Transcription factor Sp1, TSFP1
Rabbit
Antigen Affinity Chromatography
RUO
20683, 24790, 6667
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O89090, P08047, Q01714
SP1
SP1 Fusion Protein Ag16720
20 μL
Primary
Rat, Mouse, Human