missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP1 Rabbit anti-Human, Mouse, Rat, Clone: 5I3A8, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £425.00
Specifications
| Antigen | SP1 |
|---|---|
| Clone | 5I3A8 |
| Dilution | Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | ChIP Assay, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18355724
|
Novus Biologicals
NBP3-15837-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18379863
|
Novus Biologicals
NBP3-15837-100UL |
100 μg |
£425.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SP1 Monoclonal antibody specifically detects SP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| SP1 | |
| Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Sp1 transcription factor, specificity protein 1, transcription factor Sp1, TSFP1 | |
| A synthetic peptide corresponding to a sequence within amino acids 500-600 of human SP1 (P08047). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 5I3A8 | |
| ChIP Assay, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Transcription Factors and Regulators | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6667 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title