missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20460
This item is not returnable.
View return policy
Description
SP1 Polyclonal specifically detects SP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| SP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
| Sp1 transcription factor, specificity protein 1, transcription factor Sp1, TSFP1 | |
| Rabbit | |
| 81 kDa | |
| 0.1 mL | |
| Cancer, Transcription Factors and Regulators | |
| 6667 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 0.94mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Chromatin Immunoprecipitation (ChIP) | |
| P08047 | |
| SP1 | |
| Recombinant protein encompassing a sequence within the center region of human SP1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse. Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction