missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB600-233
This item is not returnable.
View return policy
Description
SP1 Polyclonal specifically detects SP1 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation Sequencing.
Specifications
| SP1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| Sp1 transcription factor, specificity protein 1, transcription factor Sp1, TSFP1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with X. laevis, X. tropicalis, Chicken, Goat, Bovine, Dog, Horse, Rabbit, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, African elephant and Northern whitecheeked gibbon. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP Assay | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunohistochemistry 1:2000 - 1:10000, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:2000 - 1:10000, Chromatin Immunoprecipitation (ChIP) 4 ug/30 ug lysate, Chromatin Immunoprecipitation Sequencing | |
| P08047 | |
| SP1 | |
| The immunogen recognized by this antibody maps to a region between residue 750 and the C-terminus (residue 785) using the numbering given in entry XP_028606.4 (GeneID 6667). | |
| 100 μL | |
| Cancer, Transcription Factors and Regulators | |
| 6667 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction