missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-232H
This item is not returnable.
View return policy
Description
SP1 Polyclonal specifically detects SP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SP1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Sp1 transcription factor, specificity protein 1, transcription factor Sp1, TSFP1 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS with No Preservative | |
| SP1 | |
| The immunogen recognized by this antibody maps to a region between residues 700 and 740 using the numbering given in entry XP_028606.4 (GeneID 6667). | |
| 100 μL | |
| Cancer, Phospho Specific, Transcription Factors and Regulators | |
| 6667 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction