missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Wilmad™ 5mm O.D. Thin Walled High-Throughput Economy NMR Tubes
Shop All SP Scienceware Products
Click to view available options
Length (Metric):
178 mm
203 mm
229 mm
Model:
Wilmad WG-5MM-ECONOMY-7-5
Wilmad WG-5MM-ECONOMY-8-5
Wilmad WG-5MM-ECONOMY-9-5
Unit Size:
Pack of 5
Description
- High quality 5mm O.D. economy sample tube
- Best O.D. tolerance in the industry and a 30% thicker wall than any competitors' product
- 100% visual and physical inspection ensures quality, including physical dimensions and surface defects
- Fit tightly in any spinner turbine
- Minimal NMR background
- Made from borosilicate glass; meets ASTM E438 Type 1 Class B standard, recognized as N51A
- Cooling/heating of these tubes may lead to breakage; for NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, refer to Wilmad Precision NMR Tubes
- Manufactured in an ISO 9001:2008 USA facility
Specifications
Specifications
| Length (Metric) | 229 mm |
| Material | Borosilicate Glass |
| Product Type | NMR Economy Sample Tube |
| Diameter (Metric) Outer | 4.947±0.019 mm |
| Field Strength | 100MHz |
| For Use With (Application) | Economy tubes are recommended only for experiments with small organic molecules (molecular weight approx.500) at ambient temperature only. |
| Includes | Tube and disposable cap |
| Length (English) | 9 in. |
| Thickness (Metric) Wall | 0.43 mm |
| Quantity | 5/Pk. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction