missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Sterile Eye Wash Solution Stations

Product Code. 10286354
Change view
Click to view available options
Color:
Natural
Height (English):
7.5 in.
Width (English):
3.56 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Height (English) Width (English) unitSize
10286354 Natural 7.5 in. 3.56 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10286354 Supplier Bel Art™ Supplier No. F248790032

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provide effective and immediate eye irrigation by outfitting your workspace with the Bel-Art™ Sterile Eye Wash Solution Station.

  • Choose from single or double stations with 1000 mL (32 oz.) bottles of sterile, isotonic, buffered eye wash solution, ready for immediate use
  • Safety-green eye wash stations can be wall-mounted for quick access when an emergency occurs
  • Include quick instruction diagrams, multi-language instructions, a polished metal mirror and four holes for mounting with screws (hardware not included)
  • Bottles have twist-off caps and a three year shelf life from date of manufacture
  • Sterile irrigation solution is helpful in rinsing away chemical contaminants and other harmful irratants and is similar in pH value to that of human tears

Specifications

Capacity (English) 33.81 oz.
Capacity (Metric) 1000 mL
Color Natural
Flow Rate 1 gpm
Height (English) 7.5 in.
Height (Metric) 19.05 cm
Includes Tamper Evident Closures
Material LDPE
Mounting Free Standing
Product Type Eyewash Refill
Sterility Sterile
Width (English) 3.56 in.
Width (Metric) 9.04 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.