missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ SP Scienceware™ Reuseable/Disposable Desiccant Cartridges
Description
- Inert, noncorrosive and nontoxic silica gel
- Clear polystyrene dish allows instant visual inspection of silica gel, which changes color from blue to pale pink as it absorbs moisture
- Highly porous membrane covers the dish, providing excellent moisture absorption
- Desiccant can be easily regenerated by heating cartridges in a 50°C (122°F) oven overnight
- For more rapid drying, cut the membrane, transfer the spent gel crystals into a glass or porcelain dish, and place in a 150°C (300°F) oven for one hour
- Individually foil sealed
- Fits Space Saver dessicator, in addition to other plastic dessicators of similar size
Order Info
Fits Bel-Art Space Saver Desiccator (Mfr. No. 420320000) and other desiccators of similar size.
Specifications
Specifications
| Diameter (Metric) Outer | 10 cm |
| Product Type | Desiccant Cartridge |
| Material | Aluminum |
| Disposable | Single-use |
| For Use With (Application) | Use analytical balance cases and precision laboratory instruments |
| Diameter (English) Outer | 3.93 in. |
Safety and Handling
Product Identifier
- Disposable Desiccant Cartridge
- Danger
- Skin sensitiser Category 1
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H317-May cause an allergic skin reaction.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: cobalt dichloride
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?