Learn More
IKA™ SP-Rack
Product Code. 17503646 Shop All IKA Products
Change view
Click to view available options
For Use With (Application):
Hanging Storage for Drying
Unit Size:
Each
Product Code. For Use With (Application) Unit Size
17503646 Hanging Storage for Drying Each
1 options
Product Code. 17503646 Supplier IKA™ Supplier No. 0020104809
  • ONLINE EXCLUSIVE PRICE
    £67.70
    £64.00 / Pack of 1
    Save £3.70
    5% Off
Estimated Shipment: 11-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SP-Rack for ROTAVISC spindles offers sufficient space for a variety of different spindles for hanging storage close to the device or for drying.
  • The stainless steel rack (AISI 304/1.4301)
  • For ROTAVISC
  • Offers sufficient space for a variety of different spindles for hanging storage close to the device or for drying.
  • Dimensions (W x H x D): 290 x 160 x 80 mm
  • Weight: 0.633 kg
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel 1.4301
For Use With (Application) Hanging Storage for Drying