missing translation for 'onlineSavingsMsg'
Learn More

SP-D Antibody (IIIH3), Novus Biologicals™

Product Code. 18360673 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360673 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360673 Supplier Novus Biologicals Supplier No. NBP311188

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SP-D Monoclonal specifically detects SP-D in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP-D
Applications Western Blot, Immunohistochemistry (Paraffin), Immunoassay
Classification Monoclonal
Clone IIIH3
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry-Paraffin, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4
Host Species Mouse
Immunogen The monoclonal antibody IIIH3 recognizes the rat surfactant protein D (SP-D).
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6441
Target Species Human, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.