missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP-D Antibody (1.7), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-41050G
This item is not returnable.
View return policy
Description
SP-D Monoclonal specifically detects SP-D in Mouse, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| SP-D | |
| Monoclonal | |
| DyLight 488 | |
| COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody detects porcine surfactant protein D (pSP-D), a 138kD member of the collectin family of proteins. Surfactant protein D is thought to play a significant role in the innate immune response against microorganisms within the lungs and at other mucosal surfaces. SP-D is a multifunctional protein which binds selectively to saccharides and lipids on microbial surfaces promoting antimicrobial responses and regulating surfactant lipid homeostasis. SP-D expression is induced in lung surfactant in response to aerogenous bacterial infection, and upon binding to bacteria, promotes phagocytosis and antigen presentation by dendritic cells. Recent studies suggest that pSP-D could serve as a serum biomarker specific for bronchopneumonia in pigs. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1.7 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| SFTPD | |
| Purified porcine surfactant protein D | |
| 0.125 mL | |
| Cell Cycle and Replication | |
| 6441 | |
| Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction