missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP-B/Surfactant Protein B Antibody (RM370), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-77431
This item is not returnable.
View return policy
Description
SP-B/Surfactant Protein B Monoclonal antibody specifically detects SP-B/Surfactant Protein B in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| SP-B/Surfactant Protein B | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol/PBS, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 6439 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| RM370 | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:500-1:1000 | |
| 18 kDa pulmonary-surfactant protein, 6 kDa protein, PSP-B, pulmonary surfactant-associated protein B, Pulmonary surfactant-associated proteolipid SPL(Phe), SFTB3, SFTP3, SMDP1, SP-B surfactant, pulmonary-associated protein B, surfactant protein B | |
| A peptide corresponding to residues near C-terminus of human SP-B/Surfactant Protein B | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction