missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SP-B/Surfactant Protein B Polyclonal specifically detects SP-B/Surfactant Protein B in Human, Mouse, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.
Specifications
Specifications
| Antigen | SP-B/Surfactant Protein B |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay |
| Classification | Polyclonal |
| Concentration | 0.1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay |
| Formulation | 0.2 um filtered solution in PBS, 0.1% BSA |
| Gene Alias | 18 kDa pulmonary-surfactant protein, 6 kDa protein, PSP-B, pulmonary surfactant-associated protein B, Pulmonary surfactant-associated proteolipid SPL(Phe), SFTB3, SFTP3, SMDP1, SP-B surfactant, pulmonary-associated protein B, surfactant protein B |
| Host Species | Rabbit |
| Immunogen | The polyclonal antibody is raised against human-based 60-amino acid synthetic SP-B polypeptide. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Ser du en mulighed for forbedring?