missing translation for 'onlineSavingsMsg'
Learn More

SP-420, MedChemExpress

Product Code. 16467208
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467208 10 mg 10mg
16459737 5 mg 5mg
16423556 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16467208 Supplier Medchem Express Supplier No. HY16912
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SP-420 is a desferrithiocin analogue with iron-clearing efficiency with ICE value of 26.7; more potent than desferrithiocin.

SP-420 is a desferrithiocin analogue with iron-clearing efficiency with ICE value of 26.7

Specifications

Chemical Name or Material SP-420
CAS 911714-45-9
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C16H21NO6S
Quantity 10 mg
Solubility Information DMSO : ≥ 38 mg/mL (106.92 mM)
SMILES O=C([C@]1(C)N=C(C2=CC=C(OCCOCCOC)C=C2O)SC1)O
Molecular Weight (g/mol) 355.41
Formula Weight 355.41
Percent Purity 99.68%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.