missing translation for 'onlineSavingsMsg'
Learn More

[SP-4-2-(1R-trans)]-(1,2-Cyclohexanediamine-N,N') Dichloridoplatinum(II) >85%, TRC

Product Code. 30357862
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357862 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357862 Supplier Toronto Research Chemicals Supplier No. TRCC987466100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichlorodiaminocyclohexaneplatinum
CAS 61848-66-6
Molecular Formula C6 H14 Cl2 N2 Pt
Quantity 100 mg
Synonym Platinum, dichloro[(1R,2R)-1,2-cyclohexanediamine-κN1,κN2]-, (SP-4-2)-, Platinum, dichloro(1,2-cyclohexanediamine-N,N')-, [SP-4-2-(1R-trans)]-, Platinum, dichloro[(1R,2R)-1,2-cyclohexanediamine-κN,κN']-, (SP-4-2)- (9CI), (SP-4-2)-Dichloro[(1R,2R)-1,2-cyclohexanediamine-κN1,κN2]platinum, 1,2-Cyclohexanediamine, platinum complex, (1R-trans)-, 1,2-Diamminecyclohexane-dichloroplatinum(II), Dichloro((1R,2R)-trans-diaminocyclohexane)platinum(II), Dichloro((R,R)-1,2-diaminocyclohexane)platinum, Dichloro(1R,2R-1,2-diaminocyclohexane)platinum, Dichloro(1R,2R-cyclohexanediammine)platinum(II), Dichloro(trans-R,R-1,2-diaminocyclohexane)platinum, Dichloro(trans-l-1,2-cyclohexanediamine)platinum(II), Dichloro[(1R,2R)-1,2-cyclohexanediamine]platinum, Dichloro[(R,R)-1,2-cyclohexanediamine]platinum, Dichloro[trans-(-)-1,2-cyclohexanediamine]platinum(II), NSC 265460, Platinum, dichloro(1,2-cyclohexanediamine-κN,κN')-, [SP-4-2-[1R-(trans)]]-, cis-Dichlorobis((1R,2R)-cyclohexanediamine)platinum, [SP-4-2-(1R-trans)]-(1,2-Cyclohexanediamine-N,N') Dichloridoplatinum(II), Platinum, dichloro[(1R,2R)-1,2-cyclohexanediamine-kappaN1,kappaN2]-, (SP-4-2)-, Platinum, dichloro[(1R,2R)-1,2-cyclohexanediamine-kappaN,kappaN']-, (SP-4-2)- (9CI), (SP-4-2)-Dichloro[(1R,2R)-1,2-cyclohexanediamine-kappaN1,kappaN2]platinum
InChI Formula InChI=1S/C6H14N2.2ClH.Pt/c7-5-3-1-2-4-6(5)8;;;/h5-6H,1-4,7-8H2;2*1H;/q;;;+2/p-2/t5-,6-;;;/m1.../s1
SMILES [Cl-][Pt+2]1([Cl-])[NH2][C@@H]2CCCC[C@H]2[NH2]1
IUPAC Name cyclohexane-1,2-diamine;dichloroplatinum
Molecular Weight (g/mol) 380.17
Formula Weight 379.0182
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.