missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Soybean Trypsin Inhibitor, ACF

Product Code. p-7217669
Encompass_Preferred
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 17179271

Brand: STEMCELL Technologies 07457

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For inhibition of trypsin activity

  • Soybean Trypsin Inhibitor, Animal Component-Free (ACF) is obtained from cultures free of animal-derived materials
  • This trypsin inhibitor consists of a single polypeptide chain with two disulfide bridges and inactivates trypsin on an equimolar basis
  • It is able to inhibit chymotrypsin to a lesser extent, but is ineffective against esterolytic, proteolytic, or elastolytic activities of porcine elastase (Vered et al.) Soybean Trypsin Inhibitor has also been shown to inhibit human and bovine thrombin, plasmin, and leukocytic proteases

Specifications

Synonym Antitrypsin; Trypsin inhibitor; Trypsin protease inhibitor
Cell Type Pancreatic Cells, T Cells, CD4+
Quantity 1 g
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.