missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Soybean Trypsin Inhibitor, ACF
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Description
- Soybean Trypsin Inhibitor, Animal Component-Free (ACF) is obtained from cultures free of animal-derived materials
- This trypsin inhibitor consists of a single polypeptide chain with two disulfide bridges and inactivates trypsin on an equimolar basis
- It is able to inhibit chymotrypsin to a lesser extent, but is ineffective against esterolytic, proteolytic, or elastolytic activities of porcine elastase (Vered et al.) Soybean Trypsin Inhibitor has also been shown to inhibit human and bovine thrombin, plasmin, and leukocytic proteases
Specifications
Specifications
| Synonym | Antitrypsin; Trypsin inhibitor; Trypsin protease inhibitor |
| Cell Type | Pancreatic Cells, T Cells, CD4+ |
| Quantity | 1 g |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction