missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Soybean GGH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117465
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
Gamma-Glutamyl hydrolase (GGH) is a homodimeric protein that catalyzes the cleavage of the gamma-glutamyl chain of folylpoly-gamma-glutamyl substrates and is a central enzyme in folyl and antifolyl poly-gamma-glutamate metabolism. The activities of the baculovirus-expressed enzyme and the enzyme from HL-60 cells were similar on methotrexate polyglutamates. GGH is expressed in a variety of human tumors and correlates with poor prognosis.
Specifications
| Soybean GGH | |
| Polyclonal | |
| Unconjugated | |
| conjugase; FGPH; folylpolyglutamate hydrolase; gamma-GH; gamma-glutamyl hydrolase; gamma-glutamyl hydrolase (conjugase, folylpolygammaglutamyl hydrolase); Gamma-Glu-X carboxypeptidase; GGH; GH | |
| Recombinant Glycine max Gamma-glutamyl hydrolase protein (22-342aa). | |
| 100 μg | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| Plant | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction