missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Soy 100

Product Code. 16239841
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 16239841

Brand: Gibco™ 215383

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Soy 100 is a highly digested soy protein peptone that is high in nucleosides and free amino acid content.

It has high concentrations of vitamins and carbohydrates, which is recommended for the cultivation of a wide variety of microbial organisms as well as varied mammalian cell culture applications.

Soy 100 can be blended with other animal origin-free peptones, such as wheat or yeast, when looking for replacements for animal origin peptones.

Soy 100 is also part of the Gibco Peptone Preview Pak, which features our new wheat, cotton, soy, and yeast peptones. These peptones provide unique nutritional profiles compared to other cell culture supplements. The Preview Pak includes five 100 gram samples*.

*The Gibco Peptone Preview Pak is non-GMP. Larger GMP packaging sizes are available.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Dehydrated Culture Media
Format Bottle, Pail
For Use With (Application) Cell Culture and Microbial Fermentation, Tissue Culture Application
pH 6.2 to 7.6
Product Line Gibco
Quantity 100 g
Regulatory Status For Research Use or Further Manufacturing. Not for diagnostic use or direct administration in to humans or animals
Remarks There are no animal sourced ingredients or animal derived reagents used in the manufacture of this product.
Source Animal origin-free
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.