missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX9 Rabbit anti-Human, Clone: SOX9/2287R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | SOX9 |
|---|---|
| Clone | SOX9/2287R |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA 1-5 μg/mL |
| Applications | ELISA, Western Blot |
Description
SOX9 Monoclonal specifically detects SOX9 in Human samples. It is validated for ELISA.Specifications
| SOX9 | |
| 0.2 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Phospho Specific, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 6662 | |
| IgG | |
| Protein A or G purified |
| SOX9/2287R | |
| ELISA 1-5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9 | |
| Recombinant human full-length SOX9 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title