Learn More
Invitrogen™ SOX9 Monoclonal Antibody (GMPR9), eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£153.00 - £394.00
Specifications
| Antigen | SOX9 |
|---|---|
| Clone | GMPR9 |
| Concentration | 0.5 mg/mL |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15547656
|
Invitrogen™
14-9765-80 |
25 μg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15557656
![]() |
Invitrogen™
14-9765-82 |
100 μg |
£394.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This GMPR9 monoclonal antibody reacts with human and mouse Sox9, a member of the Sry HMG-box (Sox) gene family of transcription factors. Like Sry, Sox9 contains a high mobility group (HMG-box) DNA-binding domain. The Sox9 DNA-binding domain recognizes the sequence AGAACAATGG, and when bound to co-factors, functions to bend the DNA. This structural change is thought to be important for chromatin remodeling and the initiation of more broad transcriptional regulation of multiple genes. Sox9 is expressed in multiple tissue types during development and its activation of downstream targets is determined by interaction with tissue-specific co-factors. During development, Sox9 expression is important for cartilage formation (mesenchymal to chondrocyte), sex determination (testes formation), neural crest formation and retinal development. Sox9 plays a role in the differentiation of multiple cell types including glial, hair, cardiac, melanocytes, pancreatic, prostate, and intestinal epithelium. Sox9 regulates expression of the following family member genes sox5, sox6, and sox10, in addition to collagen type 2 alpha, collagen 9A1, CEACAM1, and snail2. Sox9 dysregulation is involved in craniofacial defects, cleft palate, sex reversal, and some types of cancer (melanoma, lung adenocarcinoma, and breast cancer). Sox9 has also been shown to be expressed in adult stem cells in the liver, pancreas and intestine.
SOX9 has a role in sex determination and differentiation of Sertoli cells. It is involved in chondrogenesis and regulates the expression of other genes involved in chondrogenesis by acting as a transcription factor for these genes. Translocation of this gene can cause campomelic dysplasia. SOX9 is involved in the formation of testes from the indifferent fetal gonads. It is a major molecular component of the neuron-glia switch in developing spinal cord. During normal development, SOX9 allows the prostate epithelium to outgrow into the mesenchyme and then provides basal cell support for development and maintenance of the luminal epithelium. These functions of SOX9 are subverted in prostate cancer to support tumor growth and invasion. SOX9 may direct the formation of neural crest precursors and the development of a range of neural crest derivative. It has a transcriptional regulation in melanin production in cells. Northern blot analysis shows its expression on adult testis, adult heart, and fetal brain.Specifications
| SOX9 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 2010306G03Rik; AV220920; CMD1; CMPD1; mKIAA4243; SOX 9; Sox9; SOX-9; Sox9 transcription factor; SRA1; SRXX2; SRXY10; SRY (sex determining region Y)-box 9; SRY (sex determining region Y)-box 9 (campomelic dysplasia, autosomal sex-reversal); SRY (sex determining region Y)-box9; SRY (sex-determining region Y)-box 9 protein; SRY box 9; SRY-box 9; SRY-Box 9 ; SRY-box containing 9; SRY-box containing gene 9; SRY-box containing protein 9; SRY-box transcription factor 9; SRY-related HMG-box, gene 9; transcription activator Sox9; transcription factor SOX-9 | |
| SOX9 | |
| Primary | |
| 4°C | |
| SOX9 |
| GMPR9 | |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P48436, Q04887 | |
| 140586, 20682, 6662 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.